Merck
  • The preparation of hexasilsesquioxane (T6) cages by "non aqueous" hydrolysis of trichlorosilanes.
The preparation of hexasilsesquioxane (T6) cages by "non aqueous" hydrolysis of trichlorosilanes.
Chemical communications (Cambridge, England) (2003-07-05)
Alan R Bassindale, Iain A MacKinnon, Maria G Maesano, Peter G Taylor
ABSTRACT
Hexasilsesquioxane cages (T6) have been prepared from a range of alkyl and aryl trichlorosilanes using a "non aqueous" hydrolysis with dimethyl sulfoxide.
Sigma-Aldrich
Chloroplatinic acid hydrate, ~38% Pt basis
Pricing and availability is not currently available.