Merck
  • Identification of Chemotherapy-Induced Leukemic-Regenerating Cells Reveals a Transient Vulnerability of Human AML Recurrence.
Identification of Chemotherapy-Induced Leukemic-Regenerating Cells Reveals a Transient Vulnerability of Human AML Recurrence.
Cancer cell (2018-09-12)
Allison L Boyd, Lili Aslostovar, Jennifer Reid, Wendy Ye, Borko Tanasijevic, Deanna P Porras, Zoya Shapovalova, Mohammed Almakadi, Ronan Foley, Brian Leber, Anargyros Xenocostas, Mickie Bhatia
ABSTRACT
Despite successful remission induction, recurrence of acute myeloid leukemia (AML) remains a clinical obstacle thought to be caused by the retention of dormant leukemic stem cells (LSCs). Using chemotherapy-treated AML xenografts and patient samples, we have modeled patient remission and relapse kinetics to reveal that LSCs are effectively depleted via cell-cycle recruitment, leaving the origins of relapse unclear. Post-chemotherapy, in vivo characterization at the onset of disease relapse revealed a unique molecular state of leukemic-regenerating cells (LRCs) responsible for disease re-growth. LRCs are transient, can only be detected in vivo, and are molecularly distinct from therapy-naive LSCs. We demonstrate that LRC features can be used as markers of relapse and are therapeutically targetable to prevent disease recurrence.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Thioridazine hydrochloride, ≥99%