Merck
  • USP52 acts as a deubiquitinase and promotes histone chaperone ASF1A stabilization.
USP52 acts as a deubiquitinase and promotes histone chaperone ASF1A stabilization.
Nature communications (2018-03-31)
Shangda Yang, Ling Liu, Cheng Cao, Nan Song, Yuejiao Wang, Shuai Ma, Qi Zhang, Na Yu, Xiang Ding, Fuquan Yang, Shanshan Tian, Kai Zhang, Tao Sun, Jie Yang, Zhi Yao, Shaoyuan Wu, Lei Shi
ABSTRACT
Histone chaperone ASF1A has been reported to be dysregulated in multiple tumors; however, the underlying molecular mechanism that how the abundance and function of ASF1A are regulated remains unclear. Here we report that ASF1A is physically associated with USP52, which is previously identified as a pseudo-deubiquitinase. Interestingly, we demonstrate that USP52 is a bona fide ubiquitin-specific protease, and USP52 promotes ASF1A deubiquitination and stabilization. USP52-promoted ASF1A stabilization facilitates chromatin assembly and favors cell cycle progression. Additionally, we find that USP52 is overexpressed in breast carcinomas, and its level of expression correlates with that of ASF1A. Moreover, we reveal that impairment of USP52-promoted ASF1A stabilization results in growth arrest of breast cancer cells and sensitizes these cells to DNA damage. Our experiments identify USP52 as a truly protein deubiquitinase, uncover a molecular mechanism of USP52 in chromatin assembly, and reveal a potential role of USP52 in breast carcinogenesis.
Product Description
Millipore
Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-ASF1a Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.
Millipore
EZview Red Anti-HA Affinity Gel
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone CMA312, clone CMA312, from mouse
Pricing and availability is not currently available.