
Vancomycin hydrochloride from Streptomyces orientalis

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · HCl
CAS Number:
Molecular Weight:
1485.71
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Research Area
Quality Segment
Application
Mode of Action
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing