Skip to Content
Merck
Tetrahexylammonium hydrogensulfate suitable for ion pair chromatography, LiChropur™, ≥99.0% (T)

Tetrahexylammonium hydrogensulfate

Tetrahexylammonium hydrogensulfate suitable for ion pair chromatography, LiChropur™, ≥99.0% (T)
Linear Formula:
[CH3(CH2)5]4N(HSO4)
CAS Number:
Molecular Weight:
451.75
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technique