
Terbinafine Related Compound B

Synonym(s):
(2Z)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine hydrochloride
Empirical Formula (Hill Notation):
C21H25N · HCl
CAS Number:
Molecular Weight:
327.89
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2