
Sorafenib tosilate

Synonym(s):
Sorafenib tosylate, 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, 4-methylbenzenesulfonate, BAY 43-9006 tosylate salt, BAY43-9006 tosylate salt, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-methylbenzenesulfonate
Empirical Formula (Hill Notation):
C21H16ClF3N4O3·C7H8SO3
CAS Number:
Molecular Weight:
637.03
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing