Skip to Content
Merck
Sevoflurane certified reference material, pharmaceutical secondary standard

Sevoflurane

Sevoflurane certified reference material, pharmaceutical secondary standard
Synonym(s):
1,1,1,3,3,3-Hexafluoro-2-(fluoromethoxy)propane, Sevoflurane
Empirical Formula (Hill Notation):
C4H3F7O
CAS Number:
Molecular Weight:
200.05

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3