
Salmonella enterica subsp. enterica serovar Typhimurium WDCM 00031 Vitroids™

Products
Product Category
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing