
Rivastigmine Related Compound A

Synonym(s):
(+)-O,O′-Di-p-toluoyl-D-tartaric acid, Di-p-toluoyl-D-tartaric acid
Linear Formula:
[CH3C6H4CO2CH(CO2H)-]2
CAS Number:
Molecular Weight:
386.35
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2