Skip to Content
Merck
N,N-Dimethylacetamide suitable for HPLC, ≥99.9%

N,N-Dimethylacetamide

N,N-Dimethylacetamide suitable for HPLC, ≥99.9%
Synonym(s):
N,N-Dimethylacetamide, N,N-Dimethylacetamide, N,N-Dimethylacetamide solution
Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique