
Nifedipine Nitrosophenylpyridine Analog

Synonym(s):
Dimethyl 2,6-dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylate
Empirical Formula (Hill Notation):
C17H16N2O5
CAS Number:
Molecular Weight:
328.32
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2