
Naproxen Impurity N

Synonym(s):
2-Bromo-6-methoxynaphthalene
Linear Formula:
BrC10H6OCH3
CAS Number:
Molecular Weight:
237.09
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing