Skip to Content
Merck
LiChrospher® Si 60 (5 µm) LiChrospher® Si 60 (5 µm). CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.

LiChrospher® Si 60 (5 µm) HPLC Columns

LiChrospher® Si 60 (5 µm) LiChrospher® Si 60 (5 µm). CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.
Synonym(s):
Silica gel
CAS Number:

Products

Product Category
Brand
Boiling Point (°C)
Melting Point (°C)
pH Value
Product Line
Agency
USP Suitability
Separation Technique
Column Material
Particle Size (µm)
Pore Size (Å)
Technique