
Isopimpinellin

Synonym(s):
4,9-Dimethoxy-7H-furo[3,2-g][1]benzopyran-7-one, 4,9-Dimethoxyfuro[3,2-g]chromen-7-one, 5,8-Dimethoxy-6,7-furanocoumarin, 5,8-Dimethoxypsoralen, Isoimpinellin, NSC 217988, NSC 401288
Empirical Formula (Hill Notation):
C13H10O5
CAS Number:
Molecular Weight:
246.22
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing