Skip to Content
Merck
Hydrocodone solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Hydrocodone solution

Hydrocodone solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
Hydrocodone solution
Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36

Products

Brand
Feature
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique