Skip to Content
Merck
Human Insulin solution 100 μg/mL in PBS, pH 7.2, certified reference material, ampule of 100 μL, Cerilliant®

Human Insulin solution

Human Insulin solution 100 μg/mL in PBS, pH 7.2, certified reference material, ampule of 100 μL, Cerilliant®
Empirical Formula (Hill Notation):
C257H383N65O77S6
CAS Number:
Molecular Weight:
5807.57

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique