
(±)-Kavain

Synonym(s):
DL-Kavain, trans-5,6-Dihydro-4-methoxy-6-(2-phenylethenyl)-2H-pyran-2-one, DL-Kawain
Empirical Formula (Hill Notation):
C14H14O3
CAS Number:
Molecular Weight:
230.26
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing