
Danazol

Synonym(s):
17β-Hydroxy-2,4,17α-pregnadien-20-yno[2,3-d]isoxazole, 2,4,17α-Pregnadien-20-yno[2,3-d]isoxazol-17-ol
Empirical Formula (Hill Notation):
C22H27NO2
CAS Number:
Molecular Weight:
337.46
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing