
Copper(II) oxide

Synonym(s):
Cupric oxide, Copper(II) oxide
Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Particle Size (µm)
Reagent Type
Core
Special Grade
Showing 1-11 of 11
Compare
Product Number
Description
Pricing