
Cisapride monohydrate

Synonym(s):
cis-4-Amino-5-chloro-N-[1-[3-(4-fluorophenoxy)propyl]-3-methoxy-4-piperidinyl]-2-methoxybenxamide monohydrate, Acenalin, Cipril
Empirical Formula (Hill Notation):
C23H29N3O4ClF · H2O
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing