
Cilostazol Related Compound B

Synonym(s):
3,4-Dehydrocilostazol, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-2(1H)-quinolinone
Empirical Formula (Hill Notation):
C20H25N5O2
CAS Number:
Molecular Weight:
367.44
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2