
Carbidopa Related Compound A

Synonym(s):
3-O-Methylcarbidopa, (RS)-2-Hydrazino-3-(4-hydroxy-3-methoxyphenyl)-2-methylpropionic acid
Empirical Formula (Hill Notation):
C11H16N2O4
CAS Number:
Molecular Weight:
240.26
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2