
Carbamazepine Impurity B

Synonym(s):
9-Methylacridine
Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing