
Bistrifluron

Synonym(s):
1-[2-Chloro-3,5-bis(trifluoromethyl)phenyl]-3-(2,6-difluorobenzoyl)urea
Empirical Formula (Hill Notation):
C16H7ClF8N2O2
CAS Number:
Molecular Weight:
446.68
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2