
Benzene-chromium(0) tricarbonyl

Synonym(s):
Benchrotrene, Benzene-tricarbonylchromium(0)
Linear Formula:
(C6H6)Cr(CO)3
Products
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing