Adrenocorticotropic Hormone Fragment 1-24 human, rat
Synonym(s):
Tetracosactide
Empirical Formula (Hill Notation):
C136H210N40O31S
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing