7-Hydroxy-3,4-dihydro-2(1H)-quinolinone
Synonym(s):
7-Hydroxy-1,2,3,4-tetrahydro-2-quinolinone, 7-Hydroxy-3,4-dihydrocarbostyril
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing