Skip to Content
Merck
4-Methylumbelliferyl-β-D-glucuronide hydrate suitable for fluorescence, ≥99.0% (HPLC)

4-Methylumbelliferyl-β-D-glucuronide hydrate

4-Methylumbelliferyl-β-D-glucuronide hydrate suitable for fluorescence, ≥99.0% (HPLC)
Synonym(s):
4-Methylumbelliferyl-β-D-glucopyranosiduronic acid, MUG
Empirical Formula (Hill Notation):
C16H16O9 · xH2O
CAS Number:
Molecular Weight:
352.29 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment