Skip to Content
Merck
4-Bromo-1H-indazole 95%

4-Bromo-1H-indazole

4-Bromo-1H-indazole 95%
Synonym(s):
4-Bromoindazole
Empirical Formula (Hill Notation):
C7H5BrN2
CAS Number:
Molecular Weight:
197.03

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing