
3′-N-Demethyl-3′-N-formylazithromycin

Empirical Formula (Hill Notation):
C38H70N2O13
CAS Number:
Molecular Weight:
762.97
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing