
2-Hexadecanone

Synonym(s):
Methyl tetradecyl ketone
Linear Formula:
CH3(CH2)13COCH3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing