Skip to Content
Merck
2,8,9-Trimethyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane

2,8,9-Trimethyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane

2,8,9-Trimethyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
Synonym(s):
Verkade superbase
Empirical Formula (Hill Notation):
C9H21N4P
CAS Number:
Molecular Weight:
216.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing