
2′,5′-Dihydroxyacetophenone

Synonym(s):
2,5-Dihydroxyphenyl methyl ketone, 2-Acetyl-1,4-dihydroxybenzene, 2-Acetylhydroquinone, Acetylquinol, Quinacetophenone, 2′,5′-Dihydroxyacetophenone
Linear Formula:
2,5-(OH)2-C6H3COCH3
CAS Number:
Molecular Weight:
152.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing