
10-Shogaol

Synonym(s):
[10]-Shogaol, 1-(4-Hydroxy-3-methoxyphenyl)-4-tetradecen-3-one, 10-Shogaol
Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing