Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2442

Sigma-Aldrich

Anti-AGTR1 Antibody, clone 1I12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-MXRA8 Antibody, clone 1F4 ZooMAb® Rabbit Monoclonal, AT1 receptor, AT1AR, AT1BR, Angiotensin II type-1 receptor, Type-1 angiotensin II receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I12, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 41.06 kDa
observed mol wt ~52 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

rat, canine, bovine

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... AGTR1(185)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I12 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Type-1 angiotensin II receptor (AGTR1). It targets an epitope within 14 amino acids from the first extracellular domain in the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the first extracellular domain within the N-terminal region of human Type-1 angiotensin II receptor (AGTR1).

Application

Quality Control Testing

Evaluated by Western Blotting in Human liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AGTR1 in Human liver tissue lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound AGTR1 peptide with a KD of 4.0 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 Dilution of this antibody detected AGTR1 in HeLa, HepG2, and NIH3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Type-1 angiotensin II receptor (UniProt: P30556; also known as AT1AR, AT1BR, Angiotensin II type-1 receptor, AT1 receptor) is encoded by the AGTR1 (also known as AGTR1A, AGTR1B, AT2R1, AT2R1B) gene (Gene ID: 185) in human. AGTR1 is a multi-pass membrane glycoprotein of the G-protein coupled receptor family with seven transmembrane domains. It serves as a receptor for angiotensin II that acts as a key regulator of blood pressure and sodium retention by the kidney. The activated receptor couples to G- protein G(q) and thus activates phospholipase C and increases the cytosolic Ca2+ levels, which in turn triggers cellular responses such as stimulation of protein kinase C. Over-stimulation of ATGR1 leads to hypertension, cardiovascular hypertrophy, and fibrosis, whereas blocking its activity reduces blood pressure. Two subtypes of AGTR1 (AGTR1A and AGTR1B) have been described in human, mouse, and rat. They share substantial sequence homology and wide tissue distribution. AGTR1A predomina

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service