Key Documents
TMO165218
Nunc® OmniTray(TM) Single-Well Plate with 2 notched corners
polystyrene, sterile, with lid, clear, case of 60 ea
Synonym(s):
microplate, treated microplate
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
material
clear
polystyrene
sterility
sterile
packaging
case of 60 ea
manufacturer/tradename
Nunc 165218
General description
Made from clear polystyrene with standard microplate footprint, the Thermo Scientific Nunc OmniTrays are ideal for screen libraries and DNA hybridization. Available both as a standard version with 2 notched corners and as a rectangular version optimized for automation. Surface treated for cell culture or non-treated for bacterial growth and colony isolation.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries