Skip to Content
Merck
All Photos(1)

Key Documents

SML2607

Sigma-Aldrich

Clevidipine Butyrate

≥98% (HPLC)

Synonym(s):

4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid 3-methyl 5-[(1-oxobutoxy)methyl] ester, 4-(2,3-dichlorophenyl)-1,4-dihydro-2,6-dimethyl- 3,5-pyridinedicarboxylic acid methyl (1-oxobutoxy)methyl ester, H 324/38

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23Cl2NO6
CAS Number:
Molecular Weight:
456.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(cccc1C2C(=C(NC(=C2C(=O)OC)C)C)C(=O)OCOC(=O)CCC)Cl

InChI

1S/C21H23Cl2NO6/c1-5-7-15(25)29-10-30-21(27)17-12(3)24-11(2)16(20(26)28-4)18(17)13-8-6-9-14(22)19(13)23/h6,8-9,18,24H,5,7,10H2,1-4H3

InChI key

KPBZROQVTHLCDU-UHFFFAOYSA-N

Biochem/physiol Actions

Clevidipine is a short acting, third generation dihydropyridine calcium channel blocker that is highly selective for vascular L-type calcium channels. Clevidipine has no effect on myocardial contractility or cardiac conduction.
short acting, calcium channel blocker that is highly selective for vascular L-type calcium channels

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service