Skip to Content
Merck
All Photos(1)

Key Documents

SML0850

Sigma-Aldrich

PS372424 hydrochloride

≥97% (HPLC)

Synonym(s):

(3S)-N-[(1S)-4-[(Aminoiminomethyl)amino]-1-[[(cyclohexylmethyl)amino]carbonyl]butyl]-2-(1,4-dioxo-4-phenylbutyl)-1,2,3,4-tetrahydro-3-isoquinolinecarboxamide hydrochloride, (S)-N-((S)-1-((Cyclohexylmethyl)amino)-5-guanidino-1-oxopentan-2-yl)-2-(4-oxo-4-phenylbutanoyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H44N6O4 · HCl
CAS Number:
Molecular Weight:
625.20
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCC(C1=CC=CC=C1)=O)N2[C@H](C(N[C@@H](CCCNC(N)=N)C(NCC3CCCCC3)=O)=O)CC4=CC=CC=C4C2.Cl

InChI

1S/C33H44N6O4.ClH/c34-33(35)36-19-9-16-27(31(42)37-21-23-10-3-1-4-11-23)38-32(43)28-20-25-14-7-8-15-26(25)22-39(28)30(41)18-17-29(40)24-12-5-2-6-13-24;/h2,5-8,12-15,23,27-28H,1,3-4,9-11,16-22H2,(H,37,42)(H,38,43)(H4,34,35,36);1H/t27-,28-;/m0./s1

InChI key

XHGSJQNAEULTAR-DHBRAOIWSA-N

Biochem/physiol Actions

PS372424 is a highly specific agonist of human CXCR3 that potently inhibits the migration of activated T cells toward the chemokine mixture in synovial fluid from patients with active rheumatoid arthritis. PS372424 activated ERK in T cells with a similar potency and kinetic to the natural ligand CXCL11.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service