Key Documents
MTCR2700B
MTC Bio FlipTop Freezer Box With Hinged Lid
cardboard, blue, to hold, 100 x cryovials, pkg of 5 ea
Synonym(s):
cryo tube storage box, cryogenic storage, pipette holder
About This Item
UNSPSC Code:
41122806
NACRES:
NB.84
material
blue
cardboard
feature
to hold 100 x cryovials
packaging
pkg of 5 ea
manufacturer/tradename
MTC Bio R2700-B
L × W × H
133 mm × 133 mm × 51 mm
General description
FlipTop Carboard freezer box w/ attached hinged lid, 2 inch, 100-place, assorted colors, 5/pk
Recyclable cardboard, with a liquid resistant outer coating. The 81-place, 3 inch FlipTop box is suitable for tubes up to 13mm in diameter. The 100-place, 3 inch FlipTop box is suitable for tubes up to 12mm in diameter (accepts cryovials up to 4mL). Boxes are assembled complete with dividers. Each box has drain slots on the bottom and numbered grid markings in almost every conceivable location (top, bottom, sides).
Features and Benefits
  • Attached Hinged Lids
  • Foolproof identification
  • 5 1/4 x 5 1/4 x 2 inches
  • Standard size, for existing racks
  • Available in 6 colors
  • Individual or rainbow packs
  • For 81 or 100 tubes
  • 9×9 or 10×10 format
  • Alpha-numeric coding
  • Top, sides, and bottom
Legal Information
FlipTop is a trademark of MTC Bio
MTC is a trademark of MTC Bio