Skip to Content
Merck
All Photos(1)

Key Documents

DAUDI Cell Line human

85011437, human blood, Lymphoblast

Synonym(s):

DAUDI Cell Culture, Human DAUDI Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DAUDI Cell Line human, 85011437, African-American Burkitt′s lymphoma, from human blood

biological source

human blood

growth mode

Suspension

karyotype

2n = 46, diploid

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Burkitt′s lymphoma

Cell Line Description

Derived from a 16 year old male. Positive for EBNA, carries EBV markers, complement receptors, surface bound immunoglobulin and surface markers for the Fc fragment of IgG. The cells should be handled under laboratory containment level 2. Ethnicity: Black.

Application

Tumourigenicity and Virus studies: EBV used to study leukemogenesis mechanisms.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 12
D13S317: 11,12
D16S539: 10,12
D5S818: 8,13
D7S820: 8,10
THO1: 6,7
TPOX: 8,11
vWA: 15,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C. Will increase 12 fold within 10-14 days provided fresh medium is added at 48-72 hour intervals. Cell growth is slow on recovery from frozen. Resuscitate using 20% FBS and keep culture flask standing up until exponential growth is established.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service