NISTRM8785
Air particulate matter on filter media
NIST® RM 8785
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
Quality Level
manufacturer/tradename
NIST®
application(s)
industrial hygiene
format
matrix material
Related Categories
Other Notes
Reference/Informational value is provided for the following analytes.
Total carbon (C), Elemental Carbon, Organic Carbon
See certificate for values and more details at nist.gov/SRM.
Total carbon (C), Elemental Carbon, Organic Carbon
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service