Skip to Content
Merck
All Photos(1)

Key Documents

344822

Sigma-Aldrich

α1,2-Fucosidase, Xanthomonas sp.

Catalyzes the hydrolysis of all non-reducing terminal fucose that are α1,2-linked to the galactose of terminal Gal-GlcNAc disaccharide structures.

Synonym(s):

α1,2-Fucosidase, Xanthomonas sp., α-L-Fucoside Fucohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

biological source

bacterial (Xanthomonas spp.)

Quality Level

form

liquid

specific activity

≥0.4 units/mg protein
≥100 mU/mL

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

β-galactosidase, β-hexosaminidase, α- and β-mannosidase, neuraminidases, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Native α1,2-fucosidase from Xanthomon as species. Catalyzes the hydrolysis of all non-reducing terminal fucose that are α1,2-linked to the galactose of terminal Gal-GlcNAc disaccharide structures. The enzyme will not cleave fucose linked to the GlcNAc residue.
Note: 1 mU = 1 milliunit.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the release of 1.0 µmol fucose from 2ʹ-fucosyllactose per min at 37°C, pH 5.0.

Physical form

In 25 mM NaCl, 20 mM Tris-HCl, pH 7.5.

Other Notes

Noronkoski, T., and Mononen, I. 1997. Glycobiology 7, 217.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service