Skip to Content
Merck
All Photos(1)

Key Documents

1.02051

Supelco

Chemizorb® Powder

Absorbent for spilled liquids

Synonym(s):

Absorbents for liquid spillages, Spillages, absorbents for, Hazardous liquid spillages, absorbents for, Absorbant for spilled liquids

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47131905
EC Index Number:
231-545-4
NACRES:
NB.51
Pricing and availability is not currently available.

Quality Level

form

solid

particle size

<0.1 mm (>=85%)

pH

9-10 (25 °C, 100 g/L in H2O, slurry)

density

2.3 g/cm3 at 20 °C

bulk density

120 kg/m3

storage temp.

no temp limit

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Analysis Note

Identity: passes test
pH-value (10 %; water): 9 - 11
Bulk density: about 18
Particle size (< 0.1 mm): ≥ 85 %
Water absorption capacity: about 200

Legal Information

CHEMIZORB is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service