Skip to Content
Merck
All Photos(1)

Key Documents

L-063

Supelco

Lactoferrin (low Iron)

new

certified reference material, vial of 50 mg, Cerilliant®

Synonym(s):

Lactoferrin (from bovine milk)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 50 mg

manufacturer/tradename

Cerilliant®

format

neat

storage temp.

-10 to -25°C

SMILES string

S1SC[C@@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)NCC(=O)N[C@H](C(=O)N7[C@@H](CCC7)C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@@H](C1)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC

InChI

1S/C141H224N46O29S3/c1-11-77(6)111-133(212)186-112(80(9)189)134(213)183-106(130(209)184-110(76(4)5)132(211)176-97(49-32-61-161-141(155)156)120(199)170-93(45-28-57-157-137(147)148)116(195)166-78(7)113(192)180-103(136(215)216)66-82-36-16-13-17-37-82)74-219-218-73-105(182-123(202)90(42-22-25-54-142)167-114(193)87(145)65-81-34-14-12-15-35-81)129(208)173-95(47-30-59-159-139(151)152)118(197)171-96(48-31-60-160-140(153)154)122(201)178-102(68-84-70-163-89-41-21-19-39-86(84)89)127(206)174-98(51-52-108(146)190)124(203)179-101(67-83-69-162-88-40-20-18-38-85(83)88)126(205)172-94(46-29-58-158-138(149)150)119(198)175-99(53-63-217-10)125(204)169-91(43-23-26-55-143)117(196)168-92(44-24-27-56-144)121(200)177-100(64-75(2)3)115(194)164-71-109(191)165-79(8)135(214)187-62-33-50-107(187)131(210)181-104(72-188)128(207)185-111/h12-21,34-41,69-70,75-80,87,90-107,110-112,162-163,188-189H,11,22-33,42-68,71-74,142-145H2,1-10H3,(H2,146,190)(H,164,194)(H,165,191)(H,166,195)(H,167,193)(H,168,196)(H,169,204)(H,170,199)(H,171,197)(H,172,205)(H,173,208)(H,174,206)(H,175,198)(H,176,211)(H,177,200)(H,178,201)(H,179,203)(H,180,192)(H,181,210)(H,182,202)(H,183,213)(H,184,209)(H,185,207)(H,186,212)(H,215,216)(H4,147,148,157)(H4,149,150,158)(H4,151,152,159)(H4,153,154,160)(H4,155,156,161)/t77-,78-,79-,80+,87-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,110-,111-,112-/m0/s1

InChI key

PHEGMCHJJYMADA-IKGCZBKSSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service