DWK223687
WHEATON® serum vial
tube capacity (20 mL), crimp-top
Synonym(s):
223687
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
flat bottom
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
: no
packaging
case of 120 ea Shrink-wrapped modules
manufacturer/tradename
WHEATON® 223687
tube capacity
(20 mL)
tube diam.
(30.5 mm)
fitting
(Pushfit)
Looking for similar products? Visit Product Comparison Guide
General description
Serum tubing vials. Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintains the purity of the contents.
- Clear vials manufactured from low extractable borosilicate glass that conforms to USP type 1 and ASTM E 438 Type 1, Class A requirements
- Amber vials converted from Type I borosilicate glass tubing and conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Amber glass conforms to USP 1 requirements to protect light-sensitive samples
- Tubular design provides excellent clarity and dimensional consistency from vial to vial
- Specially designed bottom radius adds strength for lyophilization applications
- Lighter weight compared to molded bottles
- Shrink-wrapped modules reduce particulate contamination
- Autoclavable
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service