Skip to Content
Merck
All Photos(1)

Key Documents

C81217

Sigma-Aldrich

3-Bromo-1-phenyl-1-propene

97%

Synonym(s):

3-Phenylallyl bromide, Cinnamyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCH2Br
CAS Number:
Molecular Weight:
197.07
Beilstein:
605833
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

refractive index

n20/D 1.621 (lit.)

bp

103 °C/22 mmHg (lit.)

mp

26-29 °C (lit.)

density

1.332 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

BrC\C=C\c1ccccc1

InChI

1S/C9H9Br/c10-8-4-7-9-5-2-1-3-6-9/h1-7H,8H2/b7-4+

InChI key

RUROFEVDCUGKHD-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service