Skip to Content
Merck
All Photos(1)

Key Documents

762962

Sigma-Aldrich

Histamine-α,α,β,β-d4 dihydrochloride

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5(D)4H5N3 · 2HCl
Molecular Weight:
188.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

2-8°C

SMILES string

NC([2H])([2H])C([2H])([2H])C1=CNC=N1.Cl.Cl

InChI

1S/C5H9N3.2ClH/c6-2-1-5-3-7-4-8-5;;/h3-4H,1-2,6H2,(H,7,8);2*1H/i1D2,2D2;;

InChI key

PPZMYIBUHIPZOS-RIZDZYNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service