Skip to Content
Merck
All Photos(1)

Key Documents

616567

Sigma-Aldrich

L-Lactic acid-3,3,3-d3 solution

85 % (w/w) in H2O, ≥98 atom % D, ≥98% (CP), ≥98% (Chiral Purity, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH(OH)CO2H
CAS Number:
Molecular Weight:
93.10
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

Packaged as gram of solution

isotopic purity

≥98 atom % D

Assay

≥98% (CP)
≥98% (Chiral Purity, HPLC)

concentration

85 % (w/w) in H2O

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

SMILES string

[2H]C([2H])([2H])[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1/i1D3

InChI key

JVTAAEKCZFNVCJ-VGCLAIIRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service