Skip to Content
Merck
All Photos(1)

Key Documents

510963

Sigma-Aldrich

2-[(Trimethylsilyl)ethynyl]aniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC≡CC6H4NH2
CAS Number:
Molecular Weight:
189.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.564 (lit.)

bp

120 °C/0.1 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C#Cc1ccccc1N

InChI

1S/C11H15NSi/c1-13(2,3)9-8-10-6-4-5-7-11(10)12/h4-7H,12H2,1-3H3

InChI key

ISXSBRZDPKQYSC-UHFFFAOYSA-N

General description

2-[(Trimethylsilyl)ethynyl]aniline is a 2-ethynylaniline derivative that can be prepared by the palladium-catalyzed reaction between 2-bromonitrobenzene and trimethylsilylacetylene.

Application

2-[(Trimethylsilyl)ethynyl]aniline (2-(2-Trimethylsilylethynyl)aniline) may be used in the synthesis of:
  • N-(2-trimethylsilylethynylphenyl)methanesulfonamide
  • 8-(2-((trimethylsilyl)ethynyl)phenylamino)-7H-naphtho[1,8-bc]acridin-7-one
  • 2-phenyl-4-alkoxy quinolines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensed heteroaromatic ring systems. XIII. One-step synthesis of 2-substituted 1-methylsulfonylindoles from N-(2-halophenyl) methanesulfonamides.
Sakamoto T, et al.
Chemical & Pharmaceutical Bulletin, 36, 1305-1308 (1988)
Synthesis and Characterization of 8-(2-((Trimethylsilyl) ethynyl) phenylamino)-7H-naphtho [1,8-bc] acridin-7-one.
Li L, et al.
Synthetic Communications, 38(24), 4415-4425 (2008)
Marella A, et al.
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society, 21(1), 1-2 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service